Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084497.3 | XP_001084497.1 | 516 | 106 | ||
GenBank mRNA | JU348144.1 | - | 89 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |