Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194219.2 | NP_001181148.1 | 665 | 107 | ||
XM_001102761.3 | XP_001102761.1 | 511 | 107 | |||
XM_002799644.3 | XP_002799690.1 | 430 | 107 | |||
XM_015114743.2 | XP_014970229.1 | 599 | 107 | |||
GenBank mRNA | JU335504.1 | - | 543 | 107 | ||
JU476442.1 | - | 583 | 107 | |||
JV047683.1 | - | 601 | 107 | |||
JV690157.1 | - | 607 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |