Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042433.2 | NP_001035898.1 | 1862 | 136 | ||
XM_015121466.2 | XP_014976952.1 | 2172 | 136 | |||
GenBank mRNA | DQ644021.1 | - | 1865 | 136 | ||
EF467993.1 | - | 1900 | 136 | |||
EF467994.1 | - | 1900 | 136 | |||
JU470652.1 | - | 2069 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |