Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111877.4 | XP_001111877.2 | 569 | 131 | ||
XM_002798180.3 | XP_002798226.2 | 471 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |