Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193467.1 | NP_001180396.1 | 81 | 105 | ||
XM_015127601.2 | XP_014983087.1 | 171 | 105 | |||
XM_028841807.1 | XP_028697640.1 | 189 | 105 | |||
XM_028841808.1 | XP_028697641.1 | 189 | 105 | |||
XM_028841809.1 | XP_028697642.1 | 171 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |