Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTARC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102192.4 | XP_001102192.2 | 2685 | 121 | ||
XM_001102284.3 | XP_001102284.1 | 2630 | 121 | |||
GenBank mRNA | JU323318.1 | - | 976 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |