Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194202.2 | NP_001181131.2 | 131 | 144 | ||
XM_015125570.2 | XP_014981056.2 | 441 | 144 | |||
XM_015125571.2 | XP_014981057.2 | 454 | 144 | |||
XM_015125573.2 | XP_014981059.2 | 441 | 144 | |||
XM_028840519.1 | XP_028696352.1 | 441 | 144 | |||
GenBank mRNA | JU323753.1 | - | 367 | 144 | ||
JU472906.1 | - | 255 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |