Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087439.4 | XP_001087439.2 | 1627 | 119 | ||
GenBank mRNA | JV724005.1 | - | 1277 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |