Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194774.2 | NP_001181703.2 | 1238 | 94 | ||
XM_001111429.4 | XP_001111429.2 | 1069 | 94 | |||
XM_015111034.2 | XP_014966520.2 | 1273 | 94 | |||
XM_015111036.2 | XP_014966522.2 | 1583 | 94 | |||
XM_015111037.2 | XP_014966523.2 | 1907 | 94 | |||
XM_015111039.2 | XP_014966525.2 | 1204 | 94 | |||
XM_015111040.2 | XP_014966526.2 | 1500 | 94 | |||
XM_028830550.1 | XP_028686383.1 | 1446 | 94 | |||
XM_028830551.1 | XP_028686384.1 | 925 | 94 | |||
GenBank mRNA | JV724118.1 | - | 483 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |