Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260916.1 | NP_001247845.1 | 463 | 129 | ||
GenBank mRNA | JU322703.1 | - | 450 | 129 | ||
JV044079.1 | - | 463 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |