Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194157.2 | NP_001181086.1 | 454 | 117 | ||
XM_001100385.4 | XP_001100385.1 | 860 | 117 | |||
XM_015122452.2 | XP_014977938.1 | 2412 | 117 | |||
XM_015122453.2 | XP_014977939.1 | 515 | 117 | |||
GenBank mRNA | JU322507.1 | - | 401 | 117 | ||
JU471205.1 | - | 444 | 117 | |||
JV602054.1 | - | 447 | 117 | |||
JV704522.1 | - | 426 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |