Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105093.4 | XP_001105093.2 | 683 | 89 | ||
XM_015135314.2 | XP_014990800.1 | 683 | 89 | |||
XM_028846489.1 | XP_028702322.1 | 683 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |