Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266410.1 | NP_001253339.1 | 31 | 67 | ||
GenBank mRNA | JU332745.1 | - | 31 | 67 | ||
JU473132.1 | - | 31 | 67 | |||
JV046074.1 | - | 31 | 67 | |||
JV601896.1 | - | 58 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |