Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091712.4 | XP_001091712.3 | 586 | 128 | ||
XM_015110697.2 | XP_014966183.2 | 925 | 128 | |||
XM_015110699.2 | XP_014966185.2 | 673 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |