Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266372.1 | NP_001253301.1 | 163 | 62 | ||
XM_028832647.1 | XP_028688480.1 | 576 | 62 | |||
XM_028832648.1 | XP_028688481.1 | 468 | 62 | |||
XM_028832649.1 | XP_028688482.1 | 468 | 62 | |||
GenBank mRNA | JU333385.1 | - | 163 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |