Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001103304.4 | XP_001103304.3 | 441 | 161 | ||
XM_015146929.2 | XP_015002415.2 | 777 | 161 | |||
XM_015146930.2 | XP_015002416.2 | 828 | 161 | |||
XM_015146931.2 | XP_015002417.2 | 777 | 161 | |||
XM_015146936.2 | XP_015002422.2 | 431 | 161 | |||
GenBank mRNA | JU477874.1 | - | 830 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |