Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194261.2 | NP_001181190.2 | 593 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |