Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839467.1 | XP_028695300.1 | 122 | 110 | ||
XM_028839468.1 | XP_028695301.1 | 541 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |