Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257802.1 | NP_001244731.1 | 234 | 126 | ||
GenBank mRNA | JU322015.1 | - | 234 | 126 | ||
JV044564.1 | - | 231 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |