Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135341.2 | XP_014990827.2 | 2602 | 90 | ||
XM_015135342.2 | XP_014990828.2 | 1993 | 90 | |||
XM_028846497.1 | XP_028702330.1 | 2857 | 90 | |||
XM_028846498.1 | XP_028702331.1 | 2799 | 90 | |||
XM_028846499.1 | XP_028702332.1 | 2799 | 90 | |||
XM_028846500.1 | XP_028702333.1 | 2799 | 90 | |||
XM_028846501.1 | XP_028702334.1 | 2799 | 90 | |||
XM_028846502.1 | XP_028702335.1 | 2766 | 90 | |||
XM_028846503.1 | XP_028702336.1 | 2799 | 90 | |||
XM_028846504.1 | XP_028702337.1 | 3084 | 90 | |||
XM_028846505.1 | XP_028702338.1 | 2768 | 90 | |||
XM_028846506.1 | XP_028702339.1 | 2518 | 90 | |||
XM_028846507.1 | XP_028702340.1 | 2991 | 90 |