Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194465.2 | NP_001181394.1 | 797 | 107 | ||
GenBank mRNA | JU321540.1 | - | 585 | 107 | ||
JU473591.1 | - | 622 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |