Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194377.2 | NP_001181306.1 | 109 | 148 | ||
GenBank mRNA | JU322813.1 | - | 434 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |