Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193784.1 | NP_001180713.1 | 257 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |