Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028845022.1 | XP_028700855.1 | 4264 | 97 | ||
XM_028845027.1 | XP_028700860.1 | 3787 | 97 | |||
XM_028845028.1 | XP_028700861.1 | 3786 | 97 | |||
XM_028845029.1 | XP_028700862.1 | 4589 | 97 | |||
XM_028845030.1 | XP_028700863.1 | 3730 | 97 |
Gene Symbol: UFSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194350.3 | NP_001181279.2 | 877 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |