Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198703.2 | NP_001185632.2 | 79 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Macaca mulatta |