Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193701.2 | NP_001180630.1 | 605 | 134 | ||
GenBank mRNA | JV722312.1 | - | 491 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |