Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194473.2 | NP_001181402.2 | 364 | 86 | ||
XM_028840540.1 | XP_028696373.1 | 534 | 86 | |||
GenBank mRNA | JU333960.1 | - | 507 | 86 | ||
JV046735.1 | - | 533 | 86 | |||
JV502558.1 | - | 85 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |