Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV059051.1 | - | 266 | 82 | ||
JV649388.1 | - | 423 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |