Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THUMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265929.2 | NP_001252858.1 | 331 | 153 | ||
XM_015125658.2 | XP_014981144.1 | 452 | 153 | |||
XM_015125659.2 | XP_014981145.1 | 452 | 153 | |||
GenBank mRNA | JU335370.1 | - | 296 | 153 | ||
JV047589.1 | - | 331 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |