Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204970.2 | NP_001191899.1 | 117 | |||
GenBank mRNA | JV613588.1 | - | 188 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |