Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001103515.4 | XP_001103515.2 | 275 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta Inventoried |