Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159300.2 | NP_001152772.1 | 601 | 101 | ||
GenBank mRNA | CK231700.1 | - | 402 | 101 | ||
JV046823.1 | - | 478 | 101 | |||
JV649197.1 | - | 421 | 101 | |||
JV712955.1 | - | 467 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |