Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171189.1 | NP_001164660.1 | 189 | 94 | ||
GenBank mRNA | JU320278.1 | - | 275 | 94 | ||
JU320279.1 | - | 275 | 94 | |||
JU472738.1 | - | 265 | 94 | |||
JV043669.1 | - | 257 | 94 | |||
JV528518.1 | - | 263 | 94 | |||
JV566462.1 | - | 247 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |