Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171841.1 | NP_001165312.1 | 505 | 66 | ||
XM_015116325.2 | XP_014971811.2 | 563 | 66 | |||
GenBank mRNA | AF218046.1 | - | 505 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |