Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194228.1 | NP_001181157.1 | 475 | 113 | ||
XM_028827360.1 | XP_028683193.1 | 510 | 113 | |||
GenBank mRNA | JU325710.1 | - | 410 | 113 | ||
JV635541.1 | - | 430 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |