Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260690.1 | NP_001247619.1 | 216 | 69 | ||
GenBank mRNA | JU321736.1 | - | 216 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |