Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260803.1 | NP_001247732.1 | 776 | 98 | ||
XM_015139441.2 | XP_014994927.1 | 863 | 98 | |||
XM_015139442.2 | XP_014994928.1 | 519 | 98 | |||
XM_028848084.1 | XP_028703917.1 | 983 | 98 | |||
GenBank mRNA | JU335843.1 | - | 683 | 98 | ||
JU335844.1 | - | 683 | 98 | |||
JU335845.1 | - | 683 | 98 | |||
JU335846.1 | - | 683 | 98 | |||
JU476720.1 | - | 777 | 98 | |||
JV045811.1 | - | 776 | 98 | |||
JV590238.1 | - | 801 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |