Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYKPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260930.2 | NP_001247859.1 | 1425 | 62 | ||
XM_028849166.1 | XP_028704999.1 | 1580 | 62 | |||
XM_028849167.1 | XP_028705000.1 | 1532 | 62 | |||
XM_028849170.1 | XP_028705003.1 | 1301 | 62 | |||
GenBank mRNA | JU322930.1 | - | 1425 | 62 | ||
JU478189.1 | - | 1499 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |