Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261697.1 | NP_001248626.1 | 206 | 93 | ||
GenBank mRNA | JU320304.1 | - | 201 | 93 | ||
JU320305.1 | - | 201 | 93 | |||
JV043513.1 | - | 206 | 93 | |||
JV637803.1 | - | 208 | 93 |