Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261727.2 | NP_001248656.1 | 1175 | 61 | ||
XM_015136070.2 | XP_014991556.2 | 1791 | 61 | |||
XM_028846861.1 | XP_028702694.1 | 1791 | 61 | |||
GenBank mRNA | JV045304.1 | - | 1175 | 61 | ||
JV636656.1 | - | 1199 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |