Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265945.1 | NP_001252874.1 | 1218 | 86 | ||
XM_015123954.2 | XP_014979440.2 | 1549 | 86 | |||
XM_015123956.2 | XP_014979442.2 | 1370 | 86 | |||
XM_028838385.1 | XP_028694218.1 | 1246 | 86 | |||
XM_028838386.1 | XP_028694219.1 | 1663 | 86 | |||
GenBank mRNA | JU323098.1 | - | 1210 | 86 | ||
JU471714.1 | - | 1208 | 86 | |||
JV048291.1 | - | 1218 | 86 |