Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266011.1 | NP_001252940.1 | 192 | 84 | ||
GenBank mRNA | JU333180.1 | - | 288 | 84 | ||
JU474491.1 | - | 302 | 84 | |||
JV043780.1 | - | 192 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |