Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266069.1 | NP_001252998.1 | 253 | 72 | ||
XM_015125484.2 | XP_014980970.1 | 327 | 72 | |||
XM_015125485.2 | XP_014980971.1 | 292 | 72 | |||
XM_028840477.1 | XP_028696310.1 | 354 | 72 | |||
GenBank mRNA | JU321669.1 | - | 276 | 72 | ||
JU475384.1 | - | 219 | 72 | |||
JV045002.1 | - | 253 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |