Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266081.1 | NP_001253010.1 | 198 | 65 | ||
XM_015122628.2 | XP_014978114.1 | 726 | 65 | |||
XM_028838363.1 | XP_028694196.1 | 636 | 65 | |||
XM_028838364.1 | XP_028694197.1 | 589 | 65 | |||
GenBank mRNA | JU323503.1 | - | 198 | 65 | ||
JU323504.1 | - | 198 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |