Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032088.1 | 41 | 61 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |