Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkiras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105839.1 | NP_001099309.1 | 3 - 4 | 541 | 71 | |
XM_006247274.2 | XP_006247336.1 | 3 - 4 | 492 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |