Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006998.1 | NP_001006999.1 | 9 - 10 | 1250 | 67 | |
XM_006230787.2 | XP_006230849.1 | 10 - 11 | 1553 | 67 | ||
XM_008760132.1 | XP_008758354.1 | 9 - 10 | 1740 | 67 | ||
XM_008760134.1 | XP_008758356.1 | 9 - 10 | 1234 | 67 | ||
GenBank mRNA | BC083850.1 | - | 9 - 10 | 1250 | 7122451 | 67 |
FQ212890.1 | - | 3 - 4 | 365 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |