Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012490.2 | NP_036622.2 | 4 - 5 | 729 | 145 | |
XM_008765665.1 | XP_008763887.1 | 5 - 6 | 1926 | 145 | ||
XM_008765666.1 | XP_008763888.1 | 5 - 6 | 1451 | 145 | ||
GenBank mRNA | BC097345.1 | - | 4 - 5 | 729 | 7455117 | 145 |
X59254.1 | - | 4 - 5 | 730 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |