Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012629.1 | NP_036761.1 | 3 - 4 | 493 | 86 | |
XM_006253912.2 | XP_006253974.1 | 3 - 4 | 385 | 86 | ||
GenBank mRNA | AF022935.1 | - | 3 - 4 | 493 | 86 | |
BC166892.1 | - | 3 - 4 | 358 | 6919622 | 86 | |
BC168729.1 | - | 3 - 4 | 364 | 6922061 | 86 | |
S46794.1 | - | 2 - 3 | 229 | 86 | ||
V01249.1 | - | 3 - 4 | 358 | 86 | ||
V01250.1 | - | 2 - 3 | 273 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons |