Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012793.2 | NP_036925.2 | 1 - 2 | 235 | 121 | |
GenBank mRNA | FM043834.1 | - | 1 - 2 | 208 | 121 | |
FQ225896.1 | - | 1 - 2 | 235 | 121 | ||
FQ227114.1 | - | 1 - 2 | 235 | 121 | ||
J03588.1 | - | 1 - 2 | 233 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |